Learn More
Enzo Life Sciences L-NAME . hydrochloride (5g). CAS: 51298-62-5
SDP
Catalog No. 502007724
Encompass_Preferred
NOS inhibitor. Inhibitor of nitric oxide synthase (NOS). More soluble than NG-Nitro-L-arginine (ALX-105-001). Suppresses the release of nitric oxide (NO) from endothelial cells. Has a different effect on the superoxide generation by NOS than L-NMMA (ALX-106-001). Alternative name: NG-Nitro-L-arginine-methyl ester . HCl. Formula: C7H15N5O4 . HCl. MW: 233.2 . 36.5. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml) or methanol (25g/ml). Long Term Storage: -20°C.
Catalog No. 50-200-7724 Supplier Enzo Life Sciences Supplier No. ALX105003G005
NOS inhibitor. Inhibitor of nitric oxide synthase (NOS). More soluble than NG-Nitro-L-arginine (ALX-105-001). Suppresses the release of nitric oxide (NO) from endothelial cells. Has a different effect on the superoxide generation by NOS than L-NMMA (ALX-106-001). Alternative name: NG-Nitro-L-arginine-methyl ester . HCl. Formula: C7H15N5O4 . HCl. MW: 233.2 . 36.5. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml) or methanol (25g/ml). Long Term Storage: -20°C.