Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-NAME hydrochloride (NG-Nitroarginine methyl ester hydrochloride) | 51298-62-5 | 99.9% | 269.69 | 500 MG
SDP

Catalog No. 5000386066
Encompass_Preferred

L-NAME hydrochloride is a nitric oxide synthase (NOS) inhibitor with an IC50 of 70 μM. It acts as a precursor to the more potent NOS inhibitor L-NOARG (IC50 of 1.4 μM), making it valuable in studies of L-arginine analogues that inhibit NOS activity. Its efficacy can increase with prolonged incubation at neutral or alkaline pH, correlating with its hydrolysis to L-NOARG. This compound is also used to induce hypertension and preeclampsia models in animal studies.

  • Inhibits NOS with an IC50 of 70 μM.
  • Precursor to NOS inhibitor L-NOARG.
  • Used to induce hypertension and preeclampsia models in animal studies.
  • Appearance: solid, white to off-white.
  • Storage: -20°C, sealed, away from moisture.
  • In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-003-86066 Supplier Medchemexpress LLC Supplier No. HY18729A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-NAME hydrochloride is a nitric oxide synthase (NOS) inhibitor with an IC50 of 70 μM. It acts as a precursor to the more potent NOS inhibitor L-NOARG (IC50 of 1.4 μM), making it valuable in studies of L-arginine analogues that inhibit NOS activity. Its efficacy can increase with prolonged incubation at neutral or alkaline pH, correlating with its hydrolysis to L-NOARG. This compound is also used to induce hypertension and preeclampsia models in animal studies.

  • Inhibits NOS with an IC50 of 70 μM.
  • Precursor to NOS inhibitor L-NOARG.
  • Used to induce hypertension and preeclampsia models in animal studies.
  • Appearance: solid, white to off-white.
  • Storage: -20°C, sealed, away from moisture.
  • In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.