Learn More
Cayman Chemical L-ANAP methyl ester (hydrochloride), Purity: ≥95%, 1 mg
SDP
Catalog No. 502969099
Encompass_Preferred
An esterified form of L-ANAP; has been used in the synthesis of L-ANAP; has been used to fluorescently label proteins in vitro through genetic encoding
Catalog No. 50-296-9099 Supplier Cayman Chemical Supplier No. 154371 MG
An esterified form of L-ANAP; has been used in the synthesis of L-ANAP; has been used to fluorescently label proteins in vitro through genetic encoding