Learn More
Cayman Chemical L-ANAP methyl ester (hydrochloride), Purity: ≥95%, 5 mg
SDP
Catalog No. 502971989
Encompass_Preferred
An esterified form of L-ANAP; has been used in the synthesis of L-ANAP; has been used to fluorescently label proteins in vitro through genetic encoding
Catalog No. 50-297-1989 Supplier Cayman Chemical Supplier No. 154375 MG
An esterified form of L-ANAP; has been used in the synthesis of L-ANAP; has been used to fluorescently label proteins in vitro through genetic encoding