Learn More
Medchemexpress LLC l-Naproxen | 23979-41-1 | C14H14O3 | 1 ML
SDP
Catalog No. 5000374008
Encompass_Preferred
l-Naproxen is an enantiomer of (S)-Naproxen. It functions as an inhibitor of Cdc42 and Rac1, with EC50 values of 96 μM and 212 μM respectively, and exhibits anti-tumor activity. Additionally, l-Naproxen is classified as a nonsteroidal anti-inflammatory agent (NSAID).
  • Enantiomer of (S)-Naproxen.
  • Inhibits Cdc42 and Rac1 activity.
  • Exhibits anti-tumor activity.
  • Functions as a nonsteroidal anti-inflammatory agent (NSAID).
  • Inhibits Rac1 and Cdc42 activity in HeLa cells.
  • Inhibits migration of immortalized human ovarian cancer cells.
  • Activity is via a COX-independent mechanism.
Catalog No. 50-003-74008 Supplier Medchemexpress LLC Supplier No. HY1503110MM/1ML
l-Naproxen is an enantiomer of (S)-Naproxen. It functions as an inhibitor of Cdc42 and Rac1, with EC50 values of 96 μM and 212 μM respectively, and exhibits anti-tumor activity. Additionally, l-Naproxen is classified as a nonsteroidal anti-inflammatory agent (NSAID).
  • Enantiomer of (S)-Naproxen.
  • Inhibits Cdc42 and Rac1 activity.
  • Exhibits anti-tumor activity.
  • Functions as a nonsteroidal anti-inflammatory agent (NSAID).
  • Inhibits Rac1 and Cdc42 activity in HeLa cells.
  • Inhibits migration of immortalized human ovarian cancer cells.
  • Activity is via a COX-independent mechanism.