Learn More
Medchemexpress LLC l-Naproxen | 23979-41-1 | MFCD00870716 | C14H14O3 | 200 MG
SDP
Catalog No. 5000375489
Encompass_Preferred
l-Naproxen, also known as (R)-Naproxen, is an enantiomer of (S)-Naproxen with demonstrated anti-tumor activity. It acts as a nonsteroidal anti-inflammatory agent (NSAID) and selectively inhibits the activity of Cdc42 and Rac1. In vitro studies highlight its ability to inhibit the migration of immortalized human ovarian cancer cells through a COX-independent mechanism.
  • Inhibits Cdc42 and Rac1 activity
  • Exhibits anti-tumor properties
  • Functions as a nonsteroidal anti-inflammatory agent
  • Selectively inhibits Rac1 and Cdc42 in HeLa cells
  • Inhibits migration of human ovarian cancer cells
  • Operates via a COX-independent mechanism
Catalog No. 50-003-75489 Supplier Medchemexpress LLC Supplier No. HY15031200MG
l-Naproxen, also known as (R)-Naproxen, is an enantiomer of (S)-Naproxen with demonstrated anti-tumor activity. It acts as a nonsteroidal anti-inflammatory agent (NSAID) and selectively inhibits the activity of Cdc42 and Rac1. In vitro studies highlight its ability to inhibit the migration of immortalized human ovarian cancer cells through a COX-independent mechanism.
  • Inhibits Cdc42 and Rac1 activity
  • Exhibits anti-tumor properties
  • Functions as a nonsteroidal anti-inflammatory agent
  • Selectively inhibits Rac1 and Cdc42 in HeLa cells
  • Inhibits migration of human ovarian cancer cells
  • Operates via a COX-independent mechanism