Learn More
Enzo Life Sciences L-NIL . dihydrochloride (50mg). CAS: 159190-45-1
SDP
Supplier:  Enzo Life Sciences ALX270010M050
Encompass_Preferred
Selective inhibitor of inducible nitric oxide synthase (iNOS; NOS II). Shows anti-inflammatory activity. Alternative name: L-N6-(1-Iminoethyl)-lysine . 2HCl. Formula: C8H17N3O2 . 2HCl. MW: 187.2 . 73.0. Purity: ≥97% (TLC). Appearance: White to off-white solid. Solubility: 100mg/ml soluble in water or methanol. Long Term Storage: -20°C. Handling: Very hygroscopic. Keep under inert gas.
Catalog No. 50-200-8366