Learn More
Apexbio Technology LLC L-NIO dihydrochloride 159190-44-0 10mg
SDP
Catalog No. 5000092302
Encompass_Preferred
L-NIO dihydrochloride (CAS 159190-44-0) is a potent inhibitor of nitric oxide synthase (NOS) targeting inducible (iNOS) endothelial (eNOS) and neuronal (nNOS) isoforms with an IC50 of 65 M By blocking NOS activity L-NIO reduces nitric oxide (NO) production in activated macrophages and neutrophils which can influence both tumor cell cytotoxicity and inflammatory responses In vitro and in vivo studies demonstrate that L-NIO modulates vascular responses preventing immune complex-induced vasculitis and counteracting hypotensive effects triggered by acetylcholine and bradykinin This compound is widely utilized to investigate the role of NO in inflammation vascular regulation and immune-mediated tissue injury
Catalog No. 50-000-92302 Supplier Apexbio Technology LLC Supplier No. B634510
L-NIO dihydrochloride (CAS 159190-44-0) is a potent inhibitor of nitric oxide synthase (NOS) targeting inducible (iNOS) endothelial (eNOS) and neuronal (nNOS) isoforms with an IC50 of 65 M By blocking NOS activity L-NIO reduces nitric oxide (NO) production in activated macrophages and neutrophils which can influence both tumor cell cytotoxicity and inflammatory responses In vitro and in vivo studies demonstrate that L-NIO modulates vascular responses preventing immune complex-induced vasculitis and counteracting hypotensive effects triggered by acetylcholine and bradykinin This compound is widely utilized to investigate the role of NO in inflammation vascular regulation and immune-mediated tissue injury