Learn More
Medchemexpress LLC L-NIO dihydrochloride | 159190-44-0 | MFCD09878280 | 97.0% | 246.13 g/mol | C7H17Cl2N3O2 | 1 ML
SDP
Catalog No. 5000212052
Encompass_Preferred
L-NIO dihydrochloride is a research-grade nitric oxide synthase (NOS) inhibitor provided as a ready-to-use stock solution and as solid quantities for biochemical and in vivo studies. It is used to inhibit iNOS, eNOS, and nNOS in cellular assays and animal models where controlled NOS modulation is required.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for consistent dosing.
  • High purity suitable for research applications (97.0%).
  • Well-characterized compound with CAS registration and molecular weight data.
  • Available in both solution and solid formats to suit experimental needs.
  • Accompanied by safety and quality documents (SDS and COA) for compliance.
  • Suitable for in vitro NOS inhibition assays and in vivo ischemia models.
Catalog No. 50-002-12052 Supplier Medchemexpress LLC Supplier No. HY10098610MM 1ML
L-NIO dihydrochloride is a research-grade nitric oxide synthase (NOS) inhibitor provided as a ready-to-use stock solution and as solid quantities for biochemical and in vivo studies. It is used to inhibit iNOS, eNOS, and nNOS in cellular assays and animal models where controlled NOS modulation is required.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for consistent dosing.
  • High purity suitable for research applications (97.0%).
  • Well-characterized compound with CAS registration and molecular weight data.
  • Available in both solution and solid formats to suit experimental needs.
  • Accompanied by safety and quality documents (SDS and COA) for compliance.
  • Suitable for in vitro NOS inhibition assays and in vivo ischemia models.