Learn More
Cayman Chemical L-NIOhydrochlorIde 10mg
SDP
Catalog No. 502893829
Encompass_Preferred
L-NIO is a non-selective inhibitor of all NOS isoforms. The Ki values for nNOS (rat), eNOS (bovine), and iNOS (mouse) are 1.7, 3.9, and 3.9 µM, respectively.{6358}
Catalog No. 50-289-3829 Supplier Cayman Chemical Supplier No. 8032010 MG
L-NIO is a non-selective inhibitor of all NOS isoforms. The Ki values for nNOS (rat), eNOS (bovine), and iNOS (mouse) are 1.7, 3.9, and 3.9 µM, respectively.{6358}