Learn More
TARGETMOL CHEMICALS INC L-NMMA acetate 10MG
SDP
Catalog No. 502876258
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-NMMA acetate (Tilarginine acetate) is an inhibitor of nitric oxide synthase of all NOS isoforms (i.e. NOS1, NOS2, and NOS3). The Ki values are 0.18, 0.4, and 6 µM for nNOS, eNOS, and iNOS, respectively. Purity ≥98%
Catalog No. 50-287-6258 Supplier TARGETMOL CHEMICALS INC Supplier No. T1569710MG
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-NMMA acetate (Tilarginine acetate) is an inhibitor of nitric oxide synthase of all NOS isoforms (i.e. NOS1, NOS2, and NOS3). The Ki values are 0.18, 0.4, and 6 µM for nNOS, eNOS, and iNOS, respectively. Purity ≥98%