Learn More
Apexbio Technology LLC L-NMMA acetate 53308-83-1 50mg
SDP
Catalog No. 5000091776
Encompass_Preferred
L-NMMA acetate (CAS 53308-83-1) is a broad-spectrum inhibitor of nitric oxide synthases (NOS) exerting inhibitory effects on neuronal (nNOS) endothelial (eNOS) and inducible (iNOS) isoforms It demonstrates submicromolar to low micromolar inhibitory activity against rat nNOS human eNOS and mouse iNOS By selectively blocking NOS-mediated nitric oxide production L-NMMA acetate is widely utilized for dissecting nitric oxide-dependent signaling pathways with research applications spanning studies of tissue injury inflammatory responses and cardiovascular system functions
Catalog No. 50-000-91776 Supplier Apexbio Technology LLC Supplier No. B644450
L-NMMA acetate (CAS 53308-83-1) is a broad-spectrum inhibitor of nitric oxide synthases (NOS) exerting inhibitory effects on neuronal (nNOS) endothelial (eNOS) and inducible (iNOS) isoforms It demonstrates submicromolar to low micromolar inhibitory activity against rat nNOS human eNOS and mouse iNOS By selectively blocking NOS-mediated nitric oxide production L-NMMA acetate is widely utilized for dissecting nitric oxide-dependent signaling pathways with research applications spanning studies of tissue injury inflammatory responses and cardiovascular system functions