Learn More
Medchemexpress LLC L-NMMA acetate | 53308-83-1 | 99.0% | 248.28 | 100 MG
SDP
Catalog No. 5000388208
Encompass_Preferred
L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.
  • Functions as a nitric oxide synthase inhibitor.
  • Inhibits all NOS isoforms (NOS1, NOS2, and NOS3).
  • Demonstrates Ki values for nNOS (rat) of approximately 0.18 μM.
  • Demonstrates Ki values for eNOS (human) of approximately 0.4 μM.
  • Demonstrates Ki values for iNOS (mouse) of approximately 6 μM.
Catalog No. 50-003-88208 Supplier Medchemexpress LLC Supplier No. HY18732A100MG
L-NMMA acetate is a nitric oxide synthase inhibitor of all NOS isoforms including NOS1, NOS2, and NOS3. The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 μM, respectively.
  • Functions as a nitric oxide synthase inhibitor.
  • Inhibits all NOS isoforms (NOS1, NOS2, and NOS3).
  • Demonstrates Ki values for nNOS (rat) of approximately 0.18 μM.
  • Demonstrates Ki values for eNOS (human) of approximately 0.4 μM.
  • Demonstrates Ki values for iNOS (mouse) of approximately 6 μM.