Learn More
Medchemexpress LLC L-NMMA (acetate) | 53308-83-1 | 99.0% | 248.28 | 25 MG
SDP
Catalog No. 5000385044
Encompass_Preferred
L-NMMA acetate is a nitric oxide synthase inhibitor that targets all NOS isoforms, including NOS1, NOS2, and NOS3. The approximate Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are 0.18, 0.4, and 6 μM, respectively. This product is intended for research use only.
  • Nitric oxide synthase inhibitor.
  • Inhibits all NOS isoforms (NOS1, NOS2, NOS3).
  • Reported Ki values: nNOS (rat) ≈ 0.18 μM, eNOS (human) ≈ 0.4 μM, iNOS (mouse) ≈ 6 μM.
  • Initial source: endogenous metabolite.
Catalog No. 50-003-85044 Supplier Medchemexpress LLC Supplier No. 25MGHY18732A
L-NMMA acetate is a nitric oxide synthase inhibitor that targets all NOS isoforms, including NOS1, NOS2, and NOS3. The approximate Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are 0.18, 0.4, and 6 μM, respectively. This product is intended for research use only.
  • Nitric oxide synthase inhibitor.
  • Inhibits all NOS isoforms (NOS1, NOS2, NOS3).
  • Reported Ki values: nNOS (rat) ≈ 0.18 μM, eNOS (human) ≈ 0.4 μM, iNOS (mouse) ≈ 6 μM.
  • Initial source: endogenous metabolite.