Learn More
Medchemexpress LLC L-NMMA acetate | 53308-83-1 | MFCD00083440 | 99.0% | 248.28 | 50 MG
SDP
Catalog No. 5000388244
Encompass_Preferred
L-NMMA acetate is a nitric oxide synthase inhibitor that targets all NOS isoforms, including NOS1, NOS2, and NOS3. Its Ki values are approximately 0.18 μM for nNOS (rat), 0.4 μM for eNOS (human), and 6 μM for iNOS (mouse). This product is intended for research use only and is not sold to patients.
  • Nitric oxide synthase inhibitor of all NOS isoforms (NOS1, NOS2, and NOS3).
  • High purity of 99.0%.
  • Available in various quantities from 1 mg to 100 mg for purchase, with larger quantities available via quote.
  • Product information includes data sheet, COA, SDS, and handling instructions.
Catalog No. 50-003-88244 Supplier Medchemexpress LLC Supplier No. HY18732A50MG
L-NMMA acetate is a nitric oxide synthase inhibitor that targets all NOS isoforms, including NOS1, NOS2, and NOS3. Its Ki values are approximately 0.18 μM for nNOS (rat), 0.4 μM for eNOS (human), and 6 μM for iNOS (mouse). This product is intended for research use only and is not sold to patients.
  • Nitric oxide synthase inhibitor of all NOS isoforms (NOS1, NOS2, and NOS3).
  • High purity of 99.0%.
  • Available in various quantities from 1 mg to 100 mg for purchase, with larger quantities available via quote.
  • Product information includes data sheet, COA, SDS, and handling instructions.