Learn More
Selleck Chemical LLC L-NMMA acetate
SDP
Catalog No. 5000003605
Encompass_Preferred
L-NMMA acetate (Tilarginine Acetate Methylarginine acetate NG-Monomethyl-L-arginine acetate L-NG-monomethyl Arginine acetate) is an acetate salt form of L-NMMA L-NMMA is a nitric oxide synthase(NOS) inhibitor with an IC50 of 4 1 M for nNOS
Catalog No. 50-000-03605 Supplier Selleck Chemical LLC Supplier No. S792025MG
L-NMMA acetate (Tilarginine Acetate Methylarginine acetate NG-Monomethyl-L-arginine acetate L-NG-monomethyl Arginine acetate) is an acetate salt form of L-NMMA L-NMMA is a nitric oxide synthase(NOS) inhibitor with an IC50 of 4 1 M for nNOS