Learn More
AdipoGen L-NMMA monoacetate
SDP
Supplier:  AdipoGen AGCR13578M025
Encompass_Preferred
Chemical. CAS 53308-83-1. Formula C7H16N4O2 . CH3COOH. MW 188.2 . 60.1. Endothelium-derived relaxing factor inhibitor. Inhibits the generation of NO from arginine. Competitive, irreversible non-selective inhibitor of all three NOS I, II, and III isoforms eNOS, iNOS and nNOS. Inhibits the synthesis of nitric oxide NO in a dose-dependent and enantiospecific fashion. Useful tool to study the role and the effects of NO in cardiovascular and gastrointestinal disorders, hypertension, diabetes, septic shock, inflammation, infection, stroke and neurodegenerative disorders.
Catalog No. 50-256-2276