Learn More
AdipoGen L-NMMA . monoacetate (5 mg)
SDP
Supplier:  AdipoGen AGCR13578M005
Encompass_Preferred
Chemical. CAS: 53308-83-1. Formula: C7H16N4O2 . CH3COOH. MW: 188.2 . 60.1. Endothelium-derived relaxing factor inhibitor. Inhibits the generation of NO from arginine. Competitive, irreversible non-selective inhibitor of all three NOS I, II, and III isoforms (eNOS, iNOS and nNOS). Inhibits the synthesis of nitric oxide (NO) in a dose-dependent and enantiospecific fashion. Useful tool to study the role and the effects of NO in cardiovascular and gastrointestinal disorders, hypertension, diabetes, septic shock, inflammation, infection, stroke and neurodegenerative disorders.
Catalog No. 50-205-3944