Learn More
Enzo Life Sciences L-Norepinephrine. bitartrate. monohydrate (1g). CAS: 108341-18-0
SDP
Catalog No. 502009475
Encompass_Preferred
Adrenergic neurotransmitter. Alternative name: L-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol . C4H6O6 . H2O, L-Noradrenaline . C4H6O6 . H2O. Purity: ≥98% (Assay). Solubility: Soluble in water (46mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9475 Supplier Enzo Life Sciences Supplier No. ALX550076G001
Adrenergic neurotransmitter. Alternative name: L-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol . C4H6O6 . H2O, L-Noradrenaline . C4H6O6 . H2O. Purity: ≥98% (Assay). Solubility: Soluble in water (46mg/ml). Long Term Storage: +4°C.