Learn More
Medchemexpress LLC L-Norleucine, 6-azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 159610-89-6 | MFCD13182319 | 99.8% | 394.42 | C21H22N4O4 | 250 MG
SDP
Catalog No. 5000209276
Encompass_Preferred
N6-Diazo-L-Fmoc-lysine is an Fmoc-protected lysine derivative bearing an azide (diazo) functional group used as a click chemistry reagent for selective conjugation to alkyne- or strained alkyne-containing molecules.
  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) using DBCO or BCN reagents.
  • High purity confirmed by HPLC (99.84% in the supplied COA).
  • Available in multiple pack sizes to support synthesis and conjugation workflows.
Catalog No. 50-002-09276 Supplier Medchemexpress LLC Supplier No. 250MGHYW048205
N6-Diazo-L-Fmoc-lysine is an Fmoc-protected lysine derivative bearing an azide (diazo) functional group used as a click chemistry reagent for selective conjugation to alkyne- or strained alkyne-containing molecules.
  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) using DBCO or BCN reagents.
  • High purity confirmed by HPLC (99.84% in the supplied COA).
  • Available in multiple pack sizes to support synthesis and conjugation workflows.