Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(+)-Norvaline, 99%

Catalog No. AC226020050
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226020050 5 g Glass Bottle
AC226020010 1 g Glass Bottle
AC226020500 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226020050 Supplier Thermo Scientific Chemicals Supplier No. 226020050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Other Amino Acids (TLC) not detectable
Packaging Glass Bottle
Beilstein 04, 416
Specific Rotation +23.5° to +25.5° (20°C, 589nm) (c=10, 6N HCl)
Merck Index 15, 6805
Specific Rotation Condition 24.50 (20.00°C c=10,6 N HCl)
Quantity 5 g
Formula Weight 117.15
Percent Purity 98.5 to 101.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Norvaline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.