Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Norvaline, MP Biomedicals™

Catalog No. ICN10249501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10249501 1 g
ICN10249505 5 g
ICN10249510 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10249501 Supplier MP Biomedicals, Inc Supplier No. 0210249501
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O

Specifications

Quantity 1 g
Formula Weight 117.1
Physical Form Solid
Chemical Name or Material L-Norvaline
EINECSNumber 229-543-3
Recommended Storage Store at room Temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.