Learn More
Cayman Chemical L-Norvaline, Purity: ≥95%, CAS: 6600-40-4, 100 g
SDP
Catalog No. 502949237
Encompass_Preferred
An inhibitor of arginase and p70S6K1; inhibits arginase-mediated urea production in J774A.1 macrophage lysates at 10 mM; inhibits TNF-α-induced p70S6K1 activation in HUVECs at 20 mM; reduces microgliosis and the level of amyloid-β fibrils in the hippocampus, as well as improves learning and memory in the Morris water maze in the 3xTg mouse model of Alzheimer’s disease
Catalog No. 50-294-9237 Supplier Cayman Chemical Supplier No. 34029100 G
An inhibitor of arginase and p70S6K1; inhibits arginase-mediated urea production in J774A.1 macrophage lysates at 10 mM; inhibits TNF-α-induced p70S6K1 activation in HUVECs at 20 mM; reduces microgliosis and the level of amyloid-β fibrils in the hippocampus, as well as improves learning and memory in the Morris water maze in the 3xTg mouse model of Alzheimer’s disease