Learn More
Medchemexpress LLC L-Octanoylcarnitine | 25243-95-2 | ≥99.0% | 287.40 | 10 MG
SDP
Catalog No. 5000437855
Encompass_Preferred
L-Octanoylcarnitine is a plasma metabolite and a physiologically active form of octanoylcarnitine. It can be used for the research of breast cancer.
  • Plasma metabolite and physiologically active form of octanoylcarnitine.
  • Can be used for breast cancer research.
  • Induces H2O2 release of rat liver mitochondria (at 0.2 mM concentration in vitro).
  • Decreases mucosal and detrusor force-flow respiration and respiratory conductance in male high-fat diet (HFD) mice in vivo.
Catalog No. 50-004-37855 Supplier Medchemexpress LLC Supplier No. HY11316110MG
L-Octanoylcarnitine is a plasma metabolite and a physiologically active form of octanoylcarnitine. It can be used for the research of breast cancer.
  • Plasma metabolite and physiologically active form of octanoylcarnitine.
  • Can be used for breast cancer research.
  • Induces H2O2 release of rat liver mitochondria (at 0.2 mM concentration in vitro).
  • Decreases mucosal and detrusor force-flow respiration and respiratory conductance in male high-fat diet (HFD) mice in vivo.