Learn More
Medchemexpress LLC L-octanoylcarnitine hydrochloride | 54377-02-5 | 98.0% | 323.86 g/mol | C15H30ClNO4 | 50 MG
SDP
Catalog No. 5000285214
Encompass_Preferred
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of L-octanoylcarnitine, an endogenous plasma metabolite supplied as a research reagent for studies of fatty acid metabolism, cellular bioenergetics, and related biochemical assays. The material is intended for in vitro research and is provided with batch-specific analytical documentation for quality verification.
  • Hydrochloride salt form for improved stability and handling.
  • High purity: 98.0% (NMR).
  • Molecular formula C15H30ClNO4; molecular weight 323.86 g/mol.
  • Supplied in small research quantities, e.g., 50 MG.
  • Batch-specific COA and datasheet available for quality assurance.
  • For research use only; not for human consumption or clinical use.
Catalog No. 50-002-85214 Supplier Medchemexpress LLC Supplier No. HYW35449850MG
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of L-octanoylcarnitine, an endogenous plasma metabolite supplied as a research reagent for studies of fatty acid metabolism, cellular bioenergetics, and related biochemical assays. The material is intended for in vitro research and is provided with batch-specific analytical documentation for quality verification.
  • Hydrochloride salt form for improved stability and handling.
  • High purity: 98.0% (NMR).
  • Molecular formula C15H30ClNO4; molecular weight 323.86 g/mol.
  • Supplied in small research quantities, e.g., 50 MG.
  • Batch-specific COA and datasheet available for quality assurance.
  • For research use only; not for human consumption or clinical use.