Learn More
Medchemexpress LLC L-octanoylcarnitine hydrochloride | 54377-02-5 | MFCD00274134 | >95.0% | 323.86 g/mol | C15H30ClNO4 | 1 ML
SDP
Catalog No. 5000285109
Encompass_Preferred
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of an endogenous medium-chain acylcarnitine used in biochemical and cellular research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material for custom stock preparation; manufacturer guidance includes solubility and storage recommendations to preserve stability.
  • Ready-to-use 10 mM solution in DMSO.
  • Also available as solid for custom concentrations.
  • High solubility in DMSO (100 mg/mL) for stock preparation.
  • Storage guidance provided for powder and in-solution conditions.
  • Suitable for studies of lipid metabolism and metabolomics.
Catalog No. 50-002-85109 Supplier Medchemexpress LLC Supplier No. HYW35449810MM/1ML
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of an endogenous medium-chain acylcarnitine used in biochemical and cellular research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid material for custom stock preparation; manufacturer guidance includes solubility and storage recommendations to preserve stability.
  • Ready-to-use 10 mM solution in DMSO.
  • Also available as solid for custom concentrations.
  • High solubility in DMSO (100 mg/mL) for stock preparation.
  • Storage guidance provided for powder and in-solution conditions.
  • Suitable for studies of lipid metabolism and metabolomics.