Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-octanoylcarnitine hydrochloride | 54377-02-5 | MFCD00274134 | 98.0% | 323.86 | C15H30ClNO4 | 100 MG
SDP

Catalog No. 5000285257
Encompass_Preferred

L-Octanoylcarnitine hydrochloride is the hydrochloride salt of the endogenous metabolite octanoylcarnitine, supplied as a solid research reagent for biochemical and metabolic studies including mitochondrial H2O2 production and fatty-acid metabolism. The material is provided with analytical documentation and recommended storage conditions for laboratory use.

  • Hydrochloride salt form for enhanced solubility in aqueous media.
  • White to off-white powder suitable for analytical and biological studies.
  • High purity (98.0%) with COA and NMR documentation.
  • Molecular weight 323.86 and formula C15H30ClNO4 for accurate calculations.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Available in small lab packages including 100 mg for pilot studies.

Catalog No. 50-002-85257 Supplier Medchemexpress LLC Supplier No. HYW354498100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Octanoylcarnitine hydrochloride is the hydrochloride salt of the endogenous metabolite octanoylcarnitine, supplied as a solid research reagent for biochemical and metabolic studies including mitochondrial H2O2 production and fatty-acid metabolism. The material is provided with analytical documentation and recommended storage conditions for laboratory use.

  • Hydrochloride salt form for enhanced solubility in aqueous media.
  • White to off-white powder suitable for analytical and biological studies.
  • High purity (98.0%) with COA and NMR documentation.
  • Molecular weight 323.86 and formula C15H30ClNO4 for accurate calculations.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Available in small lab packages including 100 mg for pilot studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.