Learn More
Medchemexpress LLC L-octanoylcarnitine hydrochloride | 54377-02-5 | MFCD00274134 | 98.0% | 323.86 g/mol | C15H30ClNO4 | 10 MG
SDP
Catalog No. 5000285051
Encompass_Preferred
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of L-octanoylcarnitine, supplied as a white powder for use as an analytical standard and research reagent in studies of fatty-acid metabolism and related biochemical assays. It is commonly used to prepare stock solutions and as a reference standard in metabolomics and in vitro assay workflows.
  • Hydrochloride salt provides reliable water solubility for solution preparation.
  • Powder form enables accurate weighing and convenient storage.
  • Suitable as an analytical standard for metabolomics and LC-MS methods.
  • Supports preparation of stock solutions for cellular and biochemical assays.
  • Batch-specific certificate of analysis available for purity verification.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-85051 Supplier Medchemexpress LLC Supplier No. HYW35449810MG
L-Octanoylcarnitine hydrochloride is the hydrochloride salt of L-octanoylcarnitine, supplied as a white powder for use as an analytical standard and research reagent in studies of fatty-acid metabolism and related biochemical assays. It is commonly used to prepare stock solutions and as a reference standard in metabolomics and in vitro assay workflows.
  • Hydrochloride salt provides reliable water solubility for solution preparation.
  • Powder form enables accurate weighing and convenient storage.
  • Suitable as an analytical standard for metabolomics and LC-MS methods.
  • Supports preparation of stock solutions for cellular and biochemical assays.
  • Batch-specific certificate of analysis available for purity verification.
  • Stable when stored under recommended conditions.