Learn More
Medchemexpress LLC L-ornithine-1,2,3,4,5-13C5 hydrochloride | 943962-21-8 | 99.5% | 173.58 g/mol | C5H13ClN2O2 | 1 MG
SDP
Catalog No. 5000302053
Encompass_Preferred
L-ornithine-1,2,3,4,5-13C5 hydrochloride is a stable-isotope labeled form of L-ornithine in which all five carbon atoms are 13C-enriched. Supplied as a solid hydrochloride salt with high chemical and isotopic purity, it is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and metabolic flux studies.
  • High isotopic enrichment enabling accurate mass spectrometry quantification.
  • 99.5% chemical purity for reliable analytical performance.
  • Solid hydrochloride form suitable for analytical handling and storage.
  • Available in small quantities for tracer experiments and method development.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-02053 Supplier Medchemexpress LLC Supplier No. HYW017018S21MG
L-ornithine-1,2,3,4,5-13C5 hydrochloride is a stable-isotope labeled form of L-ornithine in which all five carbon atoms are 13C-enriched. Supplied as a solid hydrochloride salt with high chemical and isotopic purity, it is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and metabolic flux studies.
  • High isotopic enrichment enabling accurate mass spectrometry quantification.
  • 99.5% chemical purity for reliable analytical performance.
  • Solid hydrochloride form suitable for analytical handling and storage.
  • Available in small quantities for tracer experiments and method development.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).