Learn More
Medchemexpress LLC L-ornithine-1,2,3,4,5-13C5 hydrochloride | 943962-21-8 | 99.5% | 173.59 | 13C5H13ClN2O2 | 25 MG
SDP
Catalog No. 5000302944
Encompass_Preferred
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is an isotopically labeled amino acid supplied as the hydrochloride salt for use as a stable isotope standard or tracer in analytical and metabolic studies. It provides high chemical and isotopic purity in a stable, easily handled solid form suitable for LC-MS and metabolic flux applications.
  • High isotopic enrichment and chemical purity suitable for accurate quantitation.
  • Supplied as a stable, solid hydrochloride salt for easy handling and storage.
  • Compatible with LC-MS workflows and isotope dilution assays.
  • Useful for metabolic flux analysis and tracer studies.
  • Documentation includes recommended storage and supporting SDS and COA.
Catalog No. 50-003-02944 Supplier Medchemexpress LLC Supplier No. 25MGHYW017018S2
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is an isotopically labeled amino acid supplied as the hydrochloride salt for use as a stable isotope standard or tracer in analytical and metabolic studies. It provides high chemical and isotopic purity in a stable, easily handled solid form suitable for LC-MS and metabolic flux applications.
  • High isotopic enrichment and chemical purity suitable for accurate quantitation.
  • Supplied as a stable, solid hydrochloride salt for easy handling and storage.
  • Compatible with LC-MS workflows and isotope dilution assays.
  • Useful for metabolic flux analysis and tracer studies.
  • Documentation includes recommended storage and supporting SDS and COA.