Learn More
Medchemexpress LLC L-ornithine-1,2,3,4,5-13C5 hydrochloride | 943962-21-8 | 99.5% | 173.59 | 13C5H13ClN2O2 | 5 MG
SDP
Catalog No. 5000301034
Encompass_Preferred
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a 13C5-stable isotope-labeled form of L-ornithine hydrochloride supplied as a high-purity research standard for mass spectrometry and metabolic studies. Provided as a stable hydrochloride salt, it is intended for use as an internal standard or tracer in quantitative and tracer experiments.
  • 13C5 isotopic labeling for clear mass spectrometry separation.
  • Purity 99.5% suitable for analytical applications.
  • Hydrochloride salt in a stable solid form for storage and handling.
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Useful for metabolic flux and tracer experiments.
  • Available in small package sizes optimized for analytical workflows.
Catalog No. 50-003-01034 Supplier Medchemexpress LLC Supplier No. HYW017018S25MG
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a 13C5-stable isotope-labeled form of L-ornithine hydrochloride supplied as a high-purity research standard for mass spectrometry and metabolic studies. Provided as a stable hydrochloride salt, it is intended for use as an internal standard or tracer in quantitative and tracer experiments.
  • 13C5 isotopic labeling for clear mass spectrometry separation.
  • Purity 99.5% suitable for analytical applications.
  • Hydrochloride salt in a stable solid form for storage and handling.
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Useful for metabolic flux and tracer experiments.
  • Available in small package sizes optimized for analytical workflows.