Learn More
Medchemexpress LLC L-ornithine-1,2,3,4,5-13C5 hydrochloride | 943962-21-8 | 99.5% | 173.59 | 13C5H13ClN2O2 | 50 MG
SDP
Catalog No. 5000300615
Encompass_Preferred
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a stable isotope-labeled form of L-ornithine hydrochloride provided as a white to off-white solid. It is optimized for quantitative applications in metabolic studies and mass-spectrometry, offering consistent lot-to-lot performance for use as an internal standard or tracer.
  • High isotopic and chemical purity (99.5%).
  • Suitable as an internal standard for LC-MS/MS and metabolic tracing.
  • Hydrochloride salt form for improved stability and handling.
  • Available in small quantities for method development (includes 50 mg size).
  • Store sealed at 4°C to maintain stability.
Catalog No. 50-003-00615 Supplier Medchemexpress LLC Supplier No. HYW017018S250MG
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a stable isotope-labeled form of L-ornithine hydrochloride provided as a white to off-white solid. It is optimized for quantitative applications in metabolic studies and mass-spectrometry, offering consistent lot-to-lot performance for use as an internal standard or tracer.
  • High isotopic and chemical purity (99.5%).
  • Suitable as an internal standard for LC-MS/MS and metabolic tracing.
  • Hydrochloride salt form for improved stability and handling.
  • Available in small quantities for method development (includes 50 mg size).
  • Store sealed at 4°C to maintain stability.