Learn More
Medchemexpress LLC L-ornithine-1,2,3,4,5-13C5 hydrochloride | 943962-21-8 | MFCD19105630 | 99.5% | 173.59 g/mol | 13C5H13ClN2O2 | 10 MG
SDP
Catalog No. 5000301061
Encompass_Preferred
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a stable isotope-labeled form of L-ornithine supplied as the hydrochloride salt. It is provided as a white to off-white solid and is used primarily as an internal standard for mass spectrometry, metabolite quantification, and metabolic flux analysis.
  • Isotopically labeled (13C5) for accurate mass spectrometry internal standardization.
  • High purity suitable for analytical workflows.
  • Solid, easy-to-weigh form for precise sample preparation.
  • Compatible with LC-MS and GC-MS workflows.
  • Supplied with SDS and documentation for safe handling.
Catalog No. 50-003-01061 Supplier Medchemexpress LLC Supplier No. HYW017018S210MG
L-Ornithine-1,2,3,4,5-13C5 hydrochloride is a stable isotope-labeled form of L-ornithine supplied as the hydrochloride salt. It is provided as a white to off-white solid and is used primarily as an internal standard for mass spectrometry, metabolite quantification, and metabolic flux analysis.
  • Isotopically labeled (13C5) for accurate mass spectrometry internal standardization.
  • High purity suitable for analytical workflows.
  • Solid, easy-to-weigh form for precise sample preparation.
  • Compatible with LC-MS and GC-MS workflows.
  • Supplied with SDS and documentation for safe handling.