Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine 2-oxoglutarate | 5191-97-9 | MFCD00216457 | 95.0% | 278.26 g/mol | C10H18N2O7 | 5 MG
SDP

Catalog No. 5000262810
Encompass_Preferred

L-Ornithine 2-oxoglutarate is a 1:1 salt of L-ornithine and 2-oxoglutaric (α-ketoglutaric) acid supplied as a solid research reagent for biochemical and nutritional studies. It is used in metabolic and nutritional research and is provided with analytical documentation for quality assurance.

  • Solid powder suitable for laboratory use.
  • Purity 95.0%.
  • Molecular weight 278.26 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in milligram to gram packages.
  • Analytical documents available: COA, SDS, HNMR, MS.

Catalog No. 50-002-62810 Supplier Medchemexpress LLC Supplier No. HY1315145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ornithine 2-oxoglutarate is a 1:1 salt of L-ornithine and 2-oxoglutaric (α-ketoglutaric) acid supplied as a solid research reagent for biochemical and nutritional studies. It is used in metabolic and nutritional research and is provided with analytical documentation for quality assurance.

  • Solid powder suitable for laboratory use.
  • Purity 95.0%.
  • Molecular weight 278.26 g/mol.
  • Storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in milligram to gram packages.
  • Analytical documents available: COA, SDS, HNMR, MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.