Learn More
Medchemexpress LLC L-ornithine 2-oxoglutarate | 5191-97-9 | MFCD00216457 | 95.0% | C10H18N2O7 | 10MG
SDP
Catalog No. 5000261715
Encompass_Preferred
L-ornithine 2-oxoglutarate (ornithine α-ketoglutarate, OKG) is a 1:1 amino acid salt of L-ornithine and α-ketoglutarate used in nutritional and biochemical research. It is studied for effects on amino acid transport, protein synthesis, and metabolic hormone responses, and is provided as a stable solid with established chemical identifiers for laboratory use.
  • Supports amino acid transport and protein synthesis.
  • Modulates insulin and growth hormone production.
  • Suitable for nutritional supplementation and metabolic research.
  • Available in small and bulk package sizes for varied experimental needs.
  • Provided as a solid with molecular weight 278.26 g/mol.
Catalog No. 50-002-61715 Supplier Medchemexpress LLC Supplier No. HY13151410MG
L-ornithine 2-oxoglutarate (ornithine α-ketoglutarate, OKG) is a 1:1 amino acid salt of L-ornithine and α-ketoglutarate used in nutritional and biochemical research. It is studied for effects on amino acid transport, protein synthesis, and metabolic hormone responses, and is provided as a stable solid with established chemical identifiers for laboratory use.
  • Supports amino acid transport and protein synthesis.
  • Modulates insulin and growth hormone production.
  • Suitable for nutritional supplementation and metabolic research.
  • Available in small and bulk package sizes for varied experimental needs.
  • Provided as a solid with molecular weight 278.26 g/mol.