Learn More
Selleck Chemical LLC L-Ornithine S4857-100mg
SDP
Catalog No. 503111035
Encompass_Preferred
L-ornithine ((S)-2 5-Diaminopentanoic acid) has an antifatigue effect by increasing the efficiency of energy consumption and promoting the excretion of ammonia It is one of the key reactants in the urea cycle
Catalog No. 50-311-1035 Supplier Selleck Chemical LLC Supplier No. S4857100MG
L-ornithine ((S)-2 5-Diaminopentanoic acid) has an antifatigue effect by increasing the efficiency of energy consumption and promoting the excretion of ammonia It is one of the key reactants in the urea cycle