Learn More
Apexbio Technology LLC L-Ornithine 70-26-8 500mg
SDP
Catalog No. 5000089595
Encompass_Preferred
L-Ornithine (CAS 70-26-8) is a non-proteinogenic amino acid integral to the urea cycle where it facilitates the conversion of excess ammonia into urea for excretion It arises from the enzymatic cleavage of urea from arginine and subsequently participates as a precursor for the biosynthesis of citrulline and arginine L-Ornithine s role in nitrogen metabolism makes it valuable for investigating pathways of ammonia detoxification amino acid synthesis and related metabolic disorders It is widely utilized in studies focused on liver function urea cycle deficiencies and the regulation of nitrogen homeostasis in cellular and animal models
Catalog No. 50-000-89595 Supplier Apexbio Technology LLC Supplier No. B8919500
L-Ornithine (CAS 70-26-8) is a non-proteinogenic amino acid integral to the urea cycle where it facilitates the conversion of excess ammonia into urea for excretion It arises from the enzymatic cleavage of urea from arginine and subsequently participates as a precursor for the biosynthesis of citrulline and arginine L-Ornithine s role in nitrogen metabolism makes it valuable for investigating pathways of ammonia detoxification amino acid synthesis and related metabolic disorders It is widely utilized in studies focused on liver function urea cycle deficiencies and the regulation of nitrogen homeostasis in cellular and animal models