Learn More
Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | 99.9% | 170.63 | C5H11D2ClN2O2 | 5 MG
SDP
Catalog No. 5000302055
Encompass_Preferred
L-ornithine-d2 hydrochloride is the deuterium-labeled form of L-ornithine supplied for research use as an internal standard or tracer in quantitative analyses. It is provided as a high-purity, white to off-white solid hydrochloride salt tailored for analytical workflows such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity (≈99.9%).
  • Hydrochloride salt form, solid, white to off-white.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • Available in small milligram pack sizes for analytical use.
Catalog No. 50-003-02055 Supplier Medchemexpress LLC Supplier No. HYW017018S35MG
L-ornithine-d2 hydrochloride is the deuterium-labeled form of L-ornithine supplied for research use as an internal standard or tracer in quantitative analyses. It is provided as a high-purity, white to off-white solid hydrochloride salt tailored for analytical workflows such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity (≈99.9%).
  • Hydrochloride salt form, solid, white to off-white.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • Available in small milligram pack sizes for analytical use.