Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | 99.9% | 170.63 | C5H11D2ClN2O2 | 50 MG
SDP

Catalog No. 5000301081
Encompass_Preferred

L-Ornithine-d2 hydrochloride is a deuterium-labeled analog of L-ornithine hydrochloride supplied as a white to off-white solid for research use. It is used as a stable isotope tracer or internal standard in quantitative analyses by NMR, GC-MS, and LC-MS and is useful in metabolic and biochemical studies involving the urea cycle and nitrogen disposal.

  • Deuterium-labeled tracer for quantitative mass spectrometry and NMR.
  • High purity (approximately 99.9%) suitable for analytical standards.
  • Molecular formula C5H11D2ClN2O2 and molecular weight 170.63.
  • Available in multiple pack sizes to support method development and scale-up.
  • Solid form with recommended sealed storage away from moisture and light; solvent storage at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-01081 Supplier Medchemexpress LLC Supplier No. HYW017018S350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ornithine-d2 hydrochloride is a deuterium-labeled analog of L-ornithine hydrochloride supplied as a white to off-white solid for research use. It is used as a stable isotope tracer or internal standard in quantitative analyses by NMR, GC-MS, and LC-MS and is useful in metabolic and biochemical studies involving the urea cycle and nitrogen disposal.

  • Deuterium-labeled tracer for quantitative mass spectrometry and NMR.
  • High purity (approximately 99.9%) suitable for analytical standards.
  • Molecular formula C5H11D2ClN2O2 and molecular weight 170.63.
  • Available in multiple pack sizes to support method development and scale-up.
  • Solid form with recommended sealed storage away from moisture and light; solvent storage at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.