Learn More
Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | MFCD12091013 | 99.9% | 170.63 | C5H11D2ClN2O2 | 100 MG
SDP
Catalog No. 5000301269
Encompass_Preferred
L-ornithine-d2 hydrochloride is the deuterium-labeled form of L-ornithine hydrochloride supplied for research use as a white to off-white solid. It is intended for use as a stable-isotope reagent in quantitative mass spectrometry, metabolic tracing, and other analytical applications, and is provided with documentation confirming identity and purity.
  • Deuterium-labeled stable isotope suitable for analytical standards.
  • High purity for quantitative applications (99.94%).
  • White to off-white solid for convenient handling and storage.
  • Supplied with a certificate of analysis for batch-specific data.
  • Water soluble for preparation of aqueous stock solutions.
Catalog No. 50-003-01269 Supplier Medchemexpress LLC Supplier No. HYW017018S3100MG
L-ornithine-d2 hydrochloride is the deuterium-labeled form of L-ornithine hydrochloride supplied for research use as a white to off-white solid. It is intended for use as a stable-isotope reagent in quantitative mass spectrometry, metabolic tracing, and other analytical applications, and is provided with documentation confirming identity and purity.
  • Deuterium-labeled stable isotope suitable for analytical standards.
  • High purity for quantitative applications (99.94%).
  • White to off-white solid for convenient handling and storage.
  • Supplied with a certificate of analysis for batch-specific data.
  • Water soluble for preparation of aqueous stock solutions.