Learn More
Medchemexpress LLC L-ornithine-d6 hydrochloride | 347841-40-1 | >98.0% | 174.66 g/mol | C5H7D6ClN2O2 | 10 MG
SDP
Catalog No. 5000299995
Encompass_Preferred
L-Ornithine-d6 hydrochloride is the deuterium-labeled form of L-ornithine supplied as the monohydrochloride salt. It is intended for use as an isotope-labeled reference standard in metabolomics, mass spectrometry, and quantitative analytical workflows where a labeled internal standard or tracer is required.
  • Deuterium-labeled (d6) analog for isotope dilution workflows.
  • Provided as the monohydrochloride salt for stability and solubility.
  • Typical isotopic enrichment ≥98 atom % D.
  • Chemical purity ≥98%.
  • Molecular weight 174.66 g/mol.
  • Suitable as an internal standard for LC-MS and GC-MS assays.
Catalog No. 50-002-99995 Supplier Medchemexpress LLC Supplier No. HYW017018S10MG
L-Ornithine-d6 hydrochloride is the deuterium-labeled form of L-ornithine supplied as the monohydrochloride salt. It is intended for use as an isotope-labeled reference standard in metabolomics, mass spectrometry, and quantitative analytical workflows where a labeled internal standard or tracer is required.
  • Deuterium-labeled (d6) analog for isotope dilution workflows.
  • Provided as the monohydrochloride salt for stability and solubility.
  • Typical isotopic enrichment ≥98 atom % D.
  • Chemical purity ≥98%.
  • Molecular weight 174.66 g/mol.
  • Suitable as an internal standard for LC-MS and GC-MS assays.