Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine-d6 hydrochloride | 347841-40-1 | 99.2% | 174.66 | C5H7D6ClN2O2 | 1 MG
SDP

Catalog No. 5000300436
Encompass_Preferred

L-ornithine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied for research use. It is used as a stable-isotope reagent and tracer to support accurate quantitation and metabolic studies, particularly in mass spectrometry applications.

  • Deuterium-labeled (d6) form for isotope tracing and internal standard use.
  • High chemical purity: 99.2%.
  • Molecular weight 174.66 and formula C5H7D6ClN2O2 for exact mass calculations.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Supplied for research use only; not for human use.

Catalog No. 50-003-00436 Supplier Medchemexpress LLC Supplier No. HYW017018S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ornithine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied for research use. It is used as a stable-isotope reagent and tracer to support accurate quantitation and metabolic studies, particularly in mass spectrometry applications.

  • Deuterium-labeled (d6) form for isotope tracing and internal standard use.
  • High chemical purity: 99.2%.
  • Molecular weight 174.66 and formula C5H7D6ClN2O2 for exact mass calculations.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Supplied for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.