Learn More
Medchemexpress LLC L-ornithine-d6 hydrochloride | 347841-40-1 | MFCD00209770 | 99.2% | 174.66 g/mol | C5H7D6ClN2O2 | 5 MG
SDP
Catalog No. 5000301268
Encompass_Preferred
L-ornithine-d6 hydrochloride is a deuterium-labeled amino acid supplied as the hydrochloride salt for research use. It is provided as a white to off-white solid with high purity and is intended for use as a tracer or internal standard in quantitative mass spectrometry and NMR analyses.
  • Deuterium-labeled amino acid for isotope tracing and quantitation.
  • High purity (99.2%) suitable for analytical applications.
  • Hydrochloride salt, solid, water soluble (50 mg/mL with warming).
  • Applicable as internal standard for NMR, GC-MS, and LC-MS.
  • Stable when stored sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-01268 Supplier Medchemexpress LLC Supplier No. HYW017018S5MG
L-ornithine-d6 hydrochloride is a deuterium-labeled amino acid supplied as the hydrochloride salt for research use. It is provided as a white to off-white solid with high purity and is intended for use as a tracer or internal standard in quantitative mass spectrometry and NMR analyses.
  • Deuterium-labeled amino acid for isotope tracing and quantitation.
  • High purity (99.2%) suitable for analytical applications.
  • Hydrochloride salt, solid, water soluble (50 mg/mL with warming).
  • Applicable as internal standard for NMR, GC-MS, and LC-MS.
  • Stable when stored sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).