Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ornithine-d7 hydrochloride | 2483831-57-6 | 99.6% | 175.67 | C5H6D7ClN2O2 | 50 MG
SDP

Catalog No. 5000302057
Encompass_Preferred

L-Ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied as a stable-isotope standard for analytical and metabolic research. It is intended for use as an internal standard in mass spectrometry, tracer studies, and other applications requiring isotopically labeled amino acids.

  • Deuterium-labeled (d7) isotopologue for mass spectrometry and tracer experiments.
  • Provided as the hydrochloride salt for improved stability and solubility.
  • High purity suitable for analytical applications (reported ~99.6%).
  • Available in multiple small-scale quantities for research use.
  • Molecular formula C5H6D7ClN2O2; molecular weight 175.67.

Catalog No. 50-003-02057 Supplier Medchemexpress LLC Supplier No. HYW017018S450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied as a stable-isotope standard for analytical and metabolic research. It is intended for use as an internal standard in mass spectrometry, tracer studies, and other applications requiring isotopically labeled amino acids.

  • Deuterium-labeled (d7) isotopologue for mass spectrometry and tracer experiments.
  • Provided as the hydrochloride salt for improved stability and solubility.
  • High purity suitable for analytical applications (reported ~99.6%).
  • Available in multiple small-scale quantities for research use.
  • Molecular formula C5H6D7ClN2O2; molecular weight 175.67.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.