Learn More
Medchemexpress LLC L-ornithine-d7 hydrochloride | 2483831-57-6 | 99.6% | 175.67 g/mol | C5H6D7ClN2O2 | 25 MG
SDP
Catalog No. 5000303953
Encompass_Preferred
L-Ornithine-d7 hydrochloride is a deuterated (d7) form of L-ornithine supplied as the hydrochloride salt for use as an isotope-labeled reference standard in analytical workflows such as LC-MS and metabolite tracing. It is provided as a white to off-white solid with high chemical and isotopic purity, suitable for quantitative assays.
  • High isotopic labeling enabling accurate mass spectrometry quantitation.
  • High chemical purity suitable for use as a reference standard.
  • Solid, easy-to-handle powder with consistent appearance.
  • Available in multiple small pack sizes for analytical use.
  • Storage recommendations for stability: 4°C sealed; in solvent -80°C up to 6 months.
Catalog No. 50-003-03953 Supplier Medchemexpress LLC Supplier No. 25MGHYW017018S4
L-Ornithine-d7 hydrochloride is a deuterated (d7) form of L-ornithine supplied as the hydrochloride salt for use as an isotope-labeled reference standard in analytical workflows such as LC-MS and metabolite tracing. It is provided as a white to off-white solid with high chemical and isotopic purity, suitable for quantitative assays.
  • High isotopic labeling enabling accurate mass spectrometry quantitation.
  • High chemical purity suitable for use as a reference standard.
  • Solid, easy-to-handle powder with consistent appearance.
  • Available in multiple small pack sizes for analytical use.
  • Storage recommendations for stability: 4°C sealed; in solvent -80°C up to 6 months.