Learn More
Medchemexpress LLC L-ornithine-d7 hydrochloride | 2483831-57-6 | 99.9% | 175.67 | C5H6D7ClN2O2 | 1 MG
SDP
Catalog No. 5000302056
Encompass_Preferred
L-Ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of the amino acid L-ornithine, provided as a white to off-white solid. It is intended for use as an isotope-labeled reference standard and internal standard for analytical workflows such as LC-MS and stable isotope tracing in metabolomics and biochemical studies. High isotopic labeling and high purity support accurate quantitation.
  • Deuterium-labeled (d7) analog for isotope tracing and internal standard use.
  • High purity for reliable quantitative analysis.
  • Solid, easy-to-weigh form for preparation of small-volume standards.
  • Hydrochloride salt improves solubility in aqueous media.
  • Suitable for LC-MS and metabolomics workflows.
Catalog No. 50-003-02056 Supplier Medchemexpress LLC Supplier No. HYW017018S41MG
L-Ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of the amino acid L-ornithine, provided as a white to off-white solid. It is intended for use as an isotope-labeled reference standard and internal standard for analytical workflows such as LC-MS and stable isotope tracing in metabolomics and biochemical studies. High isotopic labeling and high purity support accurate quantitation.
  • Deuterium-labeled (d7) analog for isotope tracing and internal standard use.
  • High purity for reliable quantitative analysis.
  • Solid, easy-to-weigh form for preparation of small-volume standards.
  • Hydrochloride salt improves solubility in aqueous media.
  • Suitable for LC-MS and metabolomics workflows.