Learn More
Medchemexpress LLC L-ornithine-d7 hydrochloride | 2483831-57-6 | 99.9% | 175.67 | C5H6D7ClN2O2 | 5 MG
SDP
Catalog No. 5000302058
Encompass_Preferred
L-ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine used as an internal standard and reference material for analytical workflows. Supplied as a white to off-white solid, it is intended for quantitative mass spectrometry, metabolomics, and isotope tracing studies where isotopic labeling is required.
  • Deuterium-labeled internal standard for quantitative LC-MS/MS.
  • High isotopic and chemical purity (reported ≈99.9%) for accurate quantitation.
  • Hydrochloride salt form, stable solid suitable for storage and handling.
  • Useful in metabolomics, isotope tracing, and method validation workflows.
  • Available in small research-scale sizes for trace-level studies.
Catalog No. 50-003-02058 Supplier Medchemexpress LLC Supplier No. HYW017018S45MG
L-ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine used as an internal standard and reference material for analytical workflows. Supplied as a white to off-white solid, it is intended for quantitative mass spectrometry, metabolomics, and isotope tracing studies where isotopic labeling is required.
  • Deuterium-labeled internal standard for quantitative LC-MS/MS.
  • High isotopic and chemical purity (reported ≈99.9%) for accurate quantitation.
  • Hydrochloride salt form, stable solid suitable for storage and handling.
  • Useful in metabolomics, isotope tracing, and method validation workflows.
  • Available in small research-scale sizes for trace-level studies.