Learn More
Medchemexpress LLC L-ornithine-d7 hydrochloride | 2483831-57-6 | 99.9% | 175.67 g/mol | C5H6D7ClN2O2 | 10 MG
SDP
Catalog No. 5000302054
Encompass_Preferred
L-ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied as a white to off-white solid. It is intended for use as an internal standard or tracer in quantitative analytical methods (NMR, GC-MS, LC-MS) and for metabolic studies related to the urea cycle.
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High isotopic and chemical purity suitable for analytical applications.
  • Supplied as a stable solid hydrochloride for improved handling and storage.
  • Available in small preweighed quantities for trace-level assays.
  • Applicable as a tracer in metabolic and urea cycle research.
Catalog No. 50-003-02054 Supplier Medchemexpress LLC Supplier No. HYW017018S410MG
L-ornithine-d7 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied as a white to off-white solid. It is intended for use as an internal standard or tracer in quantitative analytical methods (NMR, GC-MS, LC-MS) and for metabolic studies related to the urea cycle.
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High isotopic and chemical purity suitable for analytical applications.
  • Supplied as a stable solid hydrochloride for improved handling and storage.
  • Available in small preweighed quantities for trace-level assays.
  • Applicable as a tracer in metabolic and urea cycle research.