Learn More
Alkali Scientific L Ornithine HCl (L 2 5 Diaminopentanoic Acid), 100 G
SDP
Catalog No. 5000661557
Encompass_Preferred
L-Ornithine Hydrochloride (100g) is a high-purity amino acid used primarily in cell culture applications, particularly in the growth of mammalian cells. This reagent is an essential component for synthesizing polyamines, which are crucial for cell division, growth, and differentiation. L-Ornithine is often used in culture media to support the growth of various cell types, including human and animal cells. It is widely utilized in both basic research and pharmaceutical applications. This 100g package is ideal for large-scale cell culture work in laboratories, providing a reliable source of ornithine for cell metabolism and growth studies.
Catalog No. 50-006-61557 Supplier Alkali Scientific Supplier No. ORN001CP
L-Ornithine Hydrochloride (100g) is a high-purity amino acid used primarily in cell culture applications, particularly in the growth of mammalian cells. This reagent is an essential component for synthesizing polyamines, which are crucial for cell division, growth, and differentiation. L-Ornithine is often used in culture media to support the growth of various cell types, including human and animal cells. It is widely utilized in both basic research and pharmaceutical applications. This 100g package is ideal for large-scale cell culture work in laboratories, providing a reliable source of ornithine for cell metabolism and growth studies.