Learn More
Alkali Scientific L Ornithine HCl (L 2 5 Diaminopentanoic Acid), 25 G
SDP
Catalog No. 5000660999
Encompass_Preferred
L-Ornithine Hydrochloride (25g) is a critical amino acid used in cell culture, molecular biology, and biochemistry research. This compound plays a key role in the biosynthesis of polyamines, which regulate cellular processes such as growth, differentiation, and apoptosis. L-Ornithine is commonly used in culture media to support the growth of mammalian and other eukaryotic cells. This 25g package is suitable for smaller-scale experiments, making it ideal for laboratories that require precise, small-volume additions to their cell culture protocols. It is also used in the preparation of supplements for tissue culture and metabolic studies.
Catalog No. 50-006-60999 Supplier Alkali Scientific Supplier No. ORN002CP
L-Ornithine Hydrochloride (25g) is a critical amino acid used in cell culture, molecular biology, and biochemistry research. This compound plays a key role in the biosynthesis of polyamines, which regulate cellular processes such as growth, differentiation, and apoptosis. L-Ornithine is commonly used in culture media to support the growth of mammalian and other eukaryotic cells. This 25g package is suitable for smaller-scale experiments, making it ideal for laboratories that require precise, small-volume additions to their cell culture protocols. It is also used in the preparation of supplements for tissue culture and metabolic studies.